Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococcus agalactiae Monoclonal Antibody (Strep12-445.3), QED Bioscience™

Mouse Monoclonal Antibody
$250.22 - $1002.00
Specifications
Antigen | Streptococcus agalactiae |
---|---|
Clone | Strep12-445.3 |
Concentration | 0.5-1.0 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-8131
![]() ![]() |
Leinco Technologies Inc
56002100UG |
100 μg |
Each for $250.22
|
|
|||||
50-227-8132
![]() ![]() |
Leinco Technologies Inc
56002500UG |
500 μg |
Each for $500.43
|
|
|||||
50-227-8130
![]() ![]() |
Leinco Technologies Inc
560021000UG |
1 mg |
Each for $1,002.00
|
|
|||||
Description
Streptococcus agalactiae Monoclonal antibody specifically detects Streptococcus agalactiae in Bacteria samples. It is validated for ELISASpecifications
Streptococcus agalactiae | |
0.5-1.0 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
UV-inactivated Strep. agalactiae cells isolated from mastitic milk | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
Strep12-445.3 | |
ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
s agalactiae; S. agalactiae | |
IgM | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title