Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus agalactiae Monoclonal Antibody (Strep13-504.5), QED Bioscience™
SDP

Catalog No. 502278136
Encompass
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8136 1 mg
50-227-8137 100 μg
50-227-8138 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-8136 Supplier Leinco Technologies Inc Supplier No. 560041000UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Streptococcus agalactiae Monoclonal antibody specifically detects Streptococcus agalactiae in Bacteria samples. It is validated for ELISA

Specifications

Antigen Streptococcus agalactiae
Applications ELISA
Classification Monoclonal
Clone Strep13-504.5
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias s agalactiae; S. agalactiae
Host Species Mouse
Immunogen UV-inactivated Strep. agalactiae cells isolated from mastitic milk
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.