Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococcus B Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB6001398C
Description
Streptococcus B Polyclonal antibody specifically detects Streptococcus B in Bacteria samples. It is validated for ImmunofluorescenceSpecifications
Streptococcus B | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
Beta strep, Group b streptococci, Group b streptococcus, Strep b | |
Streptococci, Group B | |
0.5 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction