Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptococcus Group A Monoclonal Antibody (HSA12-310.2), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA546103

 View more versions of this product

Catalog No. PIMA546103


Only null left
Add to Cart

Description

Description

MA1-10698 detects Streptococci Group A. MA1-10698 has been successfully used in ELISA applications. The MA1-10698 immunogen is UV-inactivated Group A Streptococcus cells.

The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Streptococcus Group A
Monoclonal
1.6 mg/mL
PBS with 0.1% sodium azide; pH 7.4
UV-inactivated Group A Streptococcus cells.
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA
HSA12-310.2
Unconjugated
Mouse
Protein G
RUO
Bacteria
Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.