Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptococcus Group A Monoclonal Antibody (HSA12-340.5), Invitrogen™

Catalog No. PIMA546109
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546109 1 mg
PIMA546118 100 μg
01-671-932 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546109 Supplier Invitrogen™ Supplier No. MA546109
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10701 detects Streptococci Group A. MA1-10701 has been successfully used in ELISA applications. The MA1-10701 immunogen is UV-inactivated Group A Streptococcus cells.

The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptococcus Group A
Applications ELISA
Classification Monoclonal
Clone HSA12-340.5
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen UV-inactivated Group A Streptococcus cells.
Purification Method Protein G
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.