Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus pneumoniae Antibody (128/390) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017614 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017614 0.025 mg
NB10064569 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017614 Supplier Novus Biologicals Supplier No. NB100645690.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Streptococcus pneumoniae Monoclonal specifically detects Streptococcus pneumoniae in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Streptococcus pneumoniae
Applications Immunocytochemistry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 128/390
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Alias Pneumococcal surface protein A, PSPA
Host Species Mouse
Immunogen Native, NCTC 10319
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity In a simple ELISA using Biogenesis Test Groups (BTGs) this is broadly reactive with S.pneumoniae. A small cross reactivity is observed with Haemophilus influenzae. Immunohistology: Reactive with isolates fixed by phenol, formalin and boiling.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.