Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptococcus pneumoniae Monoclonal Antibody (128/390)
GREENER_CHOICE

Catalog No. PIMA183478
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183478 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183478 Supplier Invitrogen™ Supplier No. MA183478
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Streptococcus pneumoniar antibody, clone 128/390 recognizes Streptococcus pneumoniae, a causative agent of respiratory disease in humans.

There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptococcus pneumoniae
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 128/390
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias Pneumococcus; s pneumoniae; S. pneumoniae
Host Species Mouse
Immunogen Streptococcus pneumoniae whole cells.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.