Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptococcus pneumoniae Monoclonal Antibody (S69S)
GREENER_CHOICE

Catalog No. PIMA518262
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518262 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518262 Supplier Invitrogen™ Supplier No. MA518262
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is reactive with the S. pneumoniae common C-polysaccharide (backbone epitope). Purity is >90%.

There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptococcus pneumoniae
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone S69S
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Pneumococcus; s pneumoniae; S. pneumoniae
Host Species Mouse
Immunogen Strep pneumoniae native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.