Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus pneumoniae Monoclonal Antibody (XI64), QED Bioscience™
SDP

Catalog No. 502277904
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7904 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7904 Supplier Leinco Technologies Inc Supplier No. 22001200UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Streptococcus pneumoniae Monoclonal antibody specifically detects Streptococcus pneumoniae in Bacteria samples. It is validated for ELISA, Western Blot

Specifications

Antigen Streptococcus pneumoniae
Applications ELISA, Western Blot
Classification Monoclonal
Clone XI64
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Pneumococcus; s pneumoniae; S. pneumoniae
Host Species Mouse
Immunogen Antigen for 22001 and 22002: heat-killed S. pneumoniae strain R36A.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.