Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptococcus pneumoniae Monoclonal Antibody (XI64)
GREENER_CHOICE

Catalog No. 01671861
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-861 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-861 Supplier Invitrogen™ Supplier No. MA110833
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10833 detects Streptococcus pneumoniae. MA1-10833 has been successfully used in ELISA and Western blot applications. The MA1-10833 immunogen is Heat-killed S. pneumoniae strain R36A.

There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptococcus pneumoniae
Applications ELISA
Classification Monoclonal
Clone XI64
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Pneumococcus; s pneumoniae; S. pneumoniae
Host Species Mouse
Immunogen Heat-killed S. pneumoniae strain R36A.
Purification Method Purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.