Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptococcus pneumoniae Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PIPA173011
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173011 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173011 Supplier Invitrogen™ Supplier No. PA173011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73011 antibody reacts with Streptococcus pneumoniae. PA1-73011 has been successfully used in immunofluorescence applications. The PA1-73011 antibody was raised against a Streptotcoccus serotype whole-cell blend (types 3, 4, 6, 7, 9, 14, 18, 19 & 23). For long term storage, aliquot and store at -20 °C, in the dark.

There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptococcus pneumoniae
Applications Immunocytochemistry
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias Pneumococcus; s pneumoniae; S. pneumoniae
Host Species Rabbit
Immunogen Streptotcoccus serotype whole-cell blend (types 3, 4, 6, 7, 9, 14, 18, 19 & 23).
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.