Learn More
Invitrogen™ Streptococcus pneumoniae Polyclonal Antibody, HRP

Description
The PA1-73012 antibody reacts with Streptococcus pneumoniae. PA1-73012 has been successfully used in ELISA, immunohistochemistry and western blot applications. The PA1-73012 antibody was raised against a Streptotcoccus serotype whole-cell blend (types 3, 4, 6, 7, 9, 14, 18, 19 & 23).
Specifications
Specifications
| Antigen | Streptococcus pneumoniae |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 10mg/mL BSA and 0.002% thimerosal |
| Gene Alias | Pneumococcus; s pneumoniae; S. pneumoniae |
| Host Species | Rabbit |
| Immunogen | Streptotcoccus serotype whole-cell blend (types 3, 4, 6, 7, 9, 14, 18, 19 & 23). |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.