Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococcus pneumoniae PS14 Antibody (5C7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274997
Description
Streptococcus pneumoniae PS14 Monoclonal specifically detects Streptococcus pneumoniae PS14 in Bacteria samples. It is validated for N/A.Specifications
| Streptococcus pneumoniae PS14 | |
| 5C7 | |
| Unconjugated | |
| Capsular Polysaccharide Serotype 14 PS14, ST-14 | |
| Streptococcus pneumoniae PS14 | |
| 0.1 mL | |
| Primary | |
| Bacteria | |
| Purified |
| Monoclonal | |
| 1 mg/ml | |
| PBS with 0.02% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Immunology, Virology Bacteria and Parasites | |
| Streptococcus pneumoniae serotype 14 capsule | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction