Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Streptococcus pyogenes speB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117551
Description
This antibody has been tested in direct-ELISA.
Important streptococcal virulence factor which cleaves human fibronectin and degrades vitronectin. Also cleaves human IL1B precursor to form biologically active IL1B. Can induce apoptosis in human monocytes and epithelial cells in vitro, and reduces phagocytic activity in monocytic cells. Thus, may play a role in bacterial colonization, invasion, and inhibition of wound healing.
Specifications
Streptococcus pyogenes speB | |
Polyclonal | |
Unconjugated | |
Exotoxin type B; SPP; Streptococcal cysteine proteinase; Streptococcus peptidase A; Streptopain | |
Recombinant Streptococcus pyogenes Streptopain protein (146-398aa). | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction