Learn More
Description
Specifications
Specifications
| Antigen | Streptococcus streptokinase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 5.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA 1:20-1:200 |
| Formulation | Phosphate Buffered Saline with 0.09% Sodium Azide |
| Host Species | Rabbit |
| Immunogen | Streptococcus streptokinase bacterial protein prepared from Lancefield strain of Streptococcus-beta hemolyticus. |
| Purification Method | Protein G purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
