Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptokinase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA185891
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA185891 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA185891 Supplier Invitrogen™ Supplier No. PA185891
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Sheep anti Streptokinase antibody recognizes intact streptokinase, a metallo-enzyme produced by various strains of Streptococcus that can liquify blood clots by converting plasminogen to plasmin.

Streptokinase is a metallo-enzyme produced by various strains of Streptococcus that can liquify blood clots by converting plasminogen to plasmin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptokinase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Sheep
Immunogen Purified, native Streptococcus streptokinase.
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.