Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Striated Muscle Myosin Heavy Chain Antibody (83B6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
| Antigen | Striated Muscle Myosin Heavy Chain |
|---|---|
| Clone | 83B6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
Description
Striated Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Striated Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Striated Muscle Myosin Heavy Chain | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cardiovascular Biology, Cellular Signaling | |
| Ascites | |
| IgG1 | |
| Ascites |
| 83B6 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Chicken, Rabbit | |
| Purified human skeletal muscle myosin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title