Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Striated Muscle Troponin I Antibody (42/25), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250309C
Description
Striated Muscle Troponin I Monoclonal specifically detects Striated Muscle Troponin I in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot.Specifications
Striated Muscle Troponin I | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Western Blot | |
42/25 | |
Western Blot | |
Skeletal muscle troponin I | |
Purified rabbit skeletal muscle troponin I | |
0.1 mL | |
Cellular Signaling | |
Specific for striated muscle troponin I. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction