Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Striated Muscle Troponin I Antibody (42/25), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250309MFV450
Description
Striated Muscle Troponin I Monoclonal antibody specifically detects Striated Muscle Troponin I in Human, Mouse, Rat, Chicken samples. It is validated for Western BlotSpecifications
| Striated Muscle Troponin I | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| 42/25 | |
| Western Blot | |
| Skeletal muscle troponin I | |
| Purified rabbit skeletal muscle troponin I | |
| 0.1 mL | |
| Cellular Signaling | |
| Human, Mouse, Rat, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction