Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Striatin 3/SG2NA Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12135R

Description
SG2NA was identified as a human cancer autoantigen and has now been implicated in vesicular trafficking and signal transduction. It is highly related to striatin and zinedin. It is a scaffolding protein that binds PP2A A and C subunits as well as the human homolog of the yeast protein, Mob1, which has been implicated in control of mitotic progression in yeast. It also binds calmodulin and appears to modulate PP2A activity.Specifications
Striatin 3/SG2NA | |
Polyclonal | |
Unconjugated | |
STRN3 | |
Cell cycle autoantigen SG2NA; cell cycle S/G2 nuclear autoantigen; cell-cycle autoantigen SG2NA; GS2NA; nuclear autoantigen; PPP2R6B; protein phosphatase 2 regulatory subunit B′′′beta; S/G2 antigen; SG2NA; SG2NA beta isoform; striatin 3; striatin, calmodulin binding protein 3; Striatin-3; striatin-3 35 kDa; striatin-3-gamma; Strn3 | |
Rabbit | |
Protein A | |
RUO | |
29966 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q13033 | |
STRN3 | |
KLH conjugated synthetic peptide derived from human Striatin 3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction