Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Striatin Antibody, Novus Biologicals™
SDP

Catalog No. p-5095407 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11074571T 0.025 mL
NB11074571 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11074571T Supplier Novus Biologicals Supplier No. NB11074571SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Striatin Polyclonal specifically detects Striatin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Striatin
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Immunoprecipitation 1:10-1:500
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. Q13033
Gene Alias MGC125642, SG2NA, striatin, striatin, calmodulin binding protein, striatin, calmodulin-binding protein
Gene Symbols STRN
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human Striatin protein (within residues 350-400). [Swiss-Prot# O43815]
Molecular Weight of Antigen 110 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6801
Test Specificity This antibody specific for Striatin. It does not cross-react with SG2NA.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.