Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STRIP1 Antibody (OTI7B8), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274408MFV500
Description
STRIP1 Monoclonal antibody specifically detects STRIP1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow CytometrySpecifications
STRIP1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
85369 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
OTI7B8 | |
Flow Cytometry | |
FAM40A, Family With Sequence Similarity 40, Member A, FAR11 Factor Arrest 11 Homolog A, FAR11 Factor Arrest 11 Homolog A (Yeast), FAR11A, Homolog Of Yeast FAR11 Protein 1, KIAA1761, Protein FAM40A, Striatin Interacting Protein 1, Striatin-Interacting Protein 1 | |
Full length human recombinant protein of human FAM40A (NP_149079) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction