Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525058
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525058 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525058 Supplier Invitrogen™ Supplier No. PA525058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STRIP2 Polyclonal Antibody for Western Blot, IHC (P)

Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the control of cell shape. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRIP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene STRIP2
Gene Accession No. Q8C9H6, Q9ULQ0
Gene Alias D330017J20Rik; FAM40B; family with sequence similarity 40, member B; FAR11 factor arrest 11 homolog B; FAR11B; homolog of yeast FAR11 protein 2; Kiaa1170; Protein FAM40B; striatin interacting protein 2; striatin-interacting protein 2; Striatin-interacting proteins 2; STRIP2; Stripb
Gene Symbols STRIP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 14-41 amino acids from the N-terminal region of human FA40B.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320609, 57464
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.