Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Striped Anti-Skid Tapes

Catalog No. 19103780 Shop All Brady Corporation Products
Click to view available options
Width (English):
1 in.
3 in.
4 in.
6 in.

For indoor or outdoor use

  • Made of rugged B-916 adhesive-backed polyester tape
  • Coated with thousands of particles of durable aluminum oxide grit
  • Bonds permanently to clean, dry floors and withstands grease and oil
  • Aids visibility of hazardous areas

Specifications

Dimensions (L x W) 60 ft. x 1 in.
Color Black, Yellow
Width (English) 1 in.
Width (Metric) 2.54 cm
Material B-916 Grit-Coated Polyester
Length (Metric) 18.2 m
Length (English) 60 ft.
Type Striped Anti-Skid Tape
Adhesive Type Permanent Pressure Sensitive Acrylic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.