Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121131
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121131 Supplier Invitrogen™ Supplier No. PA5121131
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-549, NIH/3T3, Mouse lung, Rat brain. The target is usually found in the following locations: Cell projection, Cytoplasm, Membrane, Peripheral membrane protein, dendritic spine. Immunogen sequence: HQRLLSCSAD GTLRLWNTTE VAPALSVFND TKELGIPASV DLVSSDPSHM VASFSKGYTS IFNMETQQRI LTLESNVDTT ANSSCQINRV ISHPTLPISI TAHEDRHIKF YDNNTGKLIH SMVAHLEAVT SLAVDPNGLY LMSGSHDCSI RLWNLESKTC IQEFTAHRKK FEESIHDVAF HPSKCYIASA GADALAKVFV.

Striatin is found in all mammalian cells assayed and may be involved in vesicular transport, dendrite growth, and cellular signaling. It binds Protein phosphatase 2A (PP2A) A and C subunits and calmodulin and appears to modulate PP2A activity. It also binds the human homolog of the yeast protein, Mob1, which has been implicated in mitotic progression in yeast. It is a member of a family of proteins that also includes SG2NA and Zinedin. This antibody does not cross-react with SG2NA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STRN
Gene Accession No. O43815, O55106, P70483
Gene Alias AU022939; D130055P19; PPP2R6A; protein phosphatase 2 regulatory subunit B'''alpha; SG2NA; Striatin; striatin, calmodulin binding protein; striatin, calmodulin-binding protein; STRN
Gene Symbols STRN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 581-780 of human STRN (NP_003153.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268980, 29149, 6801
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.