Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551922 Supplier Invitrogen™ Supplier No. PA551922
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YEPNVLAGTL VGHTDAVWGL AYSGIKNQLL SCSADGTVRL WNPQEKLPCI CTYNGDKKHG IPTSVDFIGC DPAHMVTSFN TGSAVIYDLE TSQSLVILSS QVDSGLQSNN HINRVVSHPT LPVTITAHED RHIKFFDNKT GKMI Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

SG2NA was identified as a human cancer autoantigen and has now been implicated in vesicular trafficking and signal transduction. It is highly related to striatin and zinedin. It is a scaffolding protein that binds PP2A A and C subunits as well as the human homolog of the yeast protein, Mob1, which has been implicated in control of mitotic progression in yeast. It also binds calmodulin and appears to modulate PP2A activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRN3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STRN3
Gene Accession No. Q13033
Gene Alias Cell cycle autoantigen SG2NA; cell cycle S/G2 nuclear autoantigen; cell-cycle autoantigen SG2NA; Gs2na; nuclear autoantigen; PPP2R6B; protein phosphatase 2 regulatory subunit B'''beta; s/G2 antigen; Sg2na; SG2NA beta isoform; striatin 3; striatin, calmodulin binding protein 3; striatin-3; striatin-3 35 kDa; striatin-3-gamma; Strn3
Gene Symbols STRN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STRN3. Recombinant protein control fragment (Product #RP-89109).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29966
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.