Learn More
Invitrogen™ STRN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588640
Description
Immunogen sequence: VRSLLGLSNS EPNGSVETKN LEQILNGGES PKQKGQEIKR SSGDVLETFN FLENADDSDE DEENDMIEGI PEGKDKHRMN KHKIGNEGLA ADLTDDPDTE EALKEFDFLV TAEDGEGAGE ARSSGDGTEW DKDDLSPTAE VWDVDQGLIS KLKEQYKKER KGKKGVKRAN RTKLYDMIAD LGDDELPHIP SGIINQSRSA STRMTDHEGA RAEEAEPITF PSGGGKSFIM GSDDVLLSVL GLGDLADLTV TNDADYSYDL P; Positive Samples: Mouse brain; Cellular Location: Cytoplasm, Membrane, Peripheral membrane protein.
SG2NA was identified as a human cancer autoantigen and has now been implicated in vesicular trafficking and signal transduction. It is highly related to striatin and zinedin. It is a scaffolding protein that binds PP2A A and C subunits as well as the human homolog of the yeast protein, Mob1, which has been implicated in control of mitotic progression in yeast. It also binds calmodulin and appears to modulate PP2A activity.
Specifications
STRN3 | |
Polyclonal | |
Unconjugated | |
STRN3 | |
Cell cycle autoantigen SG2NA; cell cycle S/G2 nuclear autoantigen; cell-cycle autoantigen SG2NA; Gs2na; nuclear autoantigen; PPP2R6B; protein phosphatase 2 regulatory subunit B'''beta; s/G2 antigen; Sg2na; SG2NA beta isoform; striatin 3; striatin, calmodulin binding protein 3; striatin-3; striatin-3 35 kDa; striatin-3-gamma; Strn3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
29966, 94186 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13033, Q9ERG2 | |
STRN3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 200-460 of human STRN3 (NP_0010773621). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.