Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578485 Supplier Invitrogen™ Supplier No. PA578485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain, rat brain Predicted Reactivity: Mouse (100%), Rat (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Binds calmodulin in a calcium dependent manner. May function as scaffolding or signaling protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRN4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Strn4
Gene Accession No. P58404, Q9NRL3
Gene Alias C80611; PPP2R6C; protein phosphatase 2 regulatory subunit B'''gamma; striatin 4; striatin, calmodulin binding protein 4; striatin-4; Strn4; Zin; zinedin
Gene Symbols Strn4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of mouse Striatin 4. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29888, 308392, 97387
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.