Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Strong Ammonia Solution, NF, 27-31%, Spectrum™ Chemical

Click to view available options
Quantity:
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
Description
Spectrum™ Chemical Strong Ammonia Solution, NF has many uses such as in topical pharmaceutical solutions destined for treating insect bites. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Quantity | 4 L |
Percent Purity | 27 to 31% |
Grade | NF |
Packaging | Poly-Coated Glass Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction