Learn More
Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F)
Description
Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay and ensure accuracy of results using the Thermo Scientific CEDIA Mitragynine (Kratom) Calibrators and Controls (CJ&F). The five calibrators (including negative) establish a standard curve that correlates the instrument signals with the calibrators' respective concentrations in ng/mL. This allows for semi-quantitative measurements. Two controls, one above and one below the cutoff (50ng/mL), validate the calibration curve. The cutoff calibrator provides a reference point for qualitative detection of positive or negative results. For Criminal Justice and Forensic use only in the USA.
- Ensures assay accuracy
- Supports qualitative and semi-quantitative measurements
- Available April 2019

Specifications
Specifications
Content And Storage | 2°C to 8°C |
Concentration | Low 37.5 ng/mL, High 62.5 ng/mL |
Detectable Analytes | Mitragynine |
Sample Type | Urine |
Type | Control |
Quantity | 2 x 5 mL |
For sale in the U.S.A. only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.