Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F)

Catalog No. p-7155965
Click to view available options
Quantity:
1 x 5 mL
2 x 5 mL
Type:
Calibrator
Control
Concentration:
100 ng/mL
20 ng/mL
200 ng/mL
50 ng/mL
Low 37.5 ng/mL, High 62.5 ng/mL
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.
This item is not returnable. View return policy
For Criminal Justice and Forensic Use Only.

Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay using the Thermo Scientific CEDIA Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F only).

Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay and ensure accuracy of results using the Thermo Scientific CEDIA Mitragynine (Kratom) Calibrators and Controls (CJ&F). The five calibrators (including negative) establish a standard curve that correlates the instrument signals with the calibrators' respective concentrations in ng/mL. This allows for semi-quantitative measurements. Two controls, one above and one below the cutoff (50ng/mL), validate the calibration curve. The cutoff calibrator provides a reference point for qualitative detection of positive or negative results. For Criminal Justice and Forensic use only in the USA.

  • Ensures assay accuracy
  • Supports qualitative and semi-quantitative measurements
  • Available April 2019
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Concentration 20 ng/mL
Detectable Analytes Mitragynine
Sample Type Urine
Type Calibrator
Quantity 1 x 5 mL

For sale in the U.S.A. only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.