Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strong Iodine Solution, 5% (w/v), USP, Spectrum™ Chemical
SDP

Catalog No. 18607036
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum

IO106

Spectrum™ Chemical Strong Iodine Solution, 5 Percent (w/v) (DEA List I Chemical), USP is used primarily as an antiseptic and disinfectant in pharmaceutical applications. All Spectrum Chemical USP products, including all USP iodine products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7553-56-2, 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

CAS Max % 0.05, 0.95
Grade USP
Packaging Amber Glass Bottle
Quantity 4 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.