Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Strontium acetate hemihydrate, 98% Catalog No.: AAA1614922
Only undefined left
Add to Cart

Strontium acetate hemihydrate, 98%

Catalog No. AAA1614922
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g

Catalog No. AAA1614922

Supplier: Thermo Scientific Chemicals A1614922

Only undefined left
Add to Cart
Only undefined left
Add to Cart
Request Bulk or Custom Formats

CAS: 14692-29-6 | Sr(OOCCH3)2·0.5H2O

PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14692-29-6
Molecular Formula Sr(OOCCH3)2·0.5H2O
MDL Number MFCD00198064
Synonym Acetic acid strontium salt

Specifications

Melting Point 150°C (in -0.5H2O)
Quantity 100 g
Merck Index 14,8836
Solubility Information Soluble in water.
Formula Weight 214.72 (205.71 Anhydrous)
Percent Purity 98%
Chemical Name or Material Strontium acetate hemihydrate
EINECSNumber 208-854-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Strontium acetate hemihydrate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.