Alfa Aesar™ Strontium acetate hemihydrate, Reagent Grade

Manufacturer: Alfa Aesar™ 1220330

Quantity 250g

 View more versions of this product

Catalog No. AA1220330

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14692-29-6
    Formula Weight 214.72 (205.71 Anhydrous)
    MDL Number MFCD00198064
    Merck Index 14,8836
    Chemical Name or Material Strontium acetate hemihydrate
    Grade Reagent Grade

    Specifications

    Formula Weight 214.72 (205.71 Anhydrous)
    Melting Point 150°C (in -0.5H2O)
    Physical Form Crystalline
    CAS 14692-29-6
    Merck Index 14,8836
    Quantity 250g
    Solubility Information Soluble in water
    TSCA TSCA
    MDL Number MFCD00198064
    Chemical Name or Material Strontium acetate hemihydrate
    Grade Reagent Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit