Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium aluminate, 99.5% (metals basis)

Catalog No. p-4407953
Click to view available options
Quantity:
25 g
100 g

CAS: 12004-37-4 | SrAl2O4

Strontium aluminate acts as a photo luminescent phosphor with long persistence of phosphorescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-37-4
Molecular Formula SrAl2O4
MDL Number MFCD00054054
Synonym strontium aluminate, strontium;oxido oxo alumane, dioxo-2,4-dioxa-1,5-dialumina-3-strontapentane, strontium aluminate,-100 mesh trace metals basis

Specifications

Physical Form Powder
Quantity 25 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Slightly soluble in water.
Formula Weight 205.58
Density 3.559
Chemical Name or Material Strontium aluminate
EINECSNumber 234-455-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.