Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium bromide hexahydrate, 99%

Catalog No. p-7026522
Click to view available options
Quantity:
100 g
500 g

CAS: 7789-53-9 | Br2H12O6Sr | 355.518 g/mol

Strontium bromide hexahydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Strontium bromide hexahydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (2850 g/L at 20°C).

Notes
m.p. 88° (rapidly heated) , 180° -6H{2}O
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-53-9
Molecular Formula Br2H12O6Sr
Molecular Weight (g/mol) 355.518
MDL Number MFCD00150361
InChI Key FLMJUJXBFKFYOZ-UHFFFAOYSA-L
Synonym unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
PubChem CID 165644
IUPAC Name strontium;dibromide;hexahydrate
SMILES O.O.O.O.O.O.[Br-].[Br-].[Sr+2]

Specifications

Quantity 100 g
Linear Formula SrBr2·6H2O
Merck Index 14,8837
Solubility Information Soluble in water (2850g/L at 20°C).
Formula Weight 355.53 (247.44 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.386
Chemical Name or Material Strontium bromide hexahydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-969-5
RTECSNumber WK8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.