Strontium carbonate, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 14343A1

Quantity 1kg

 View more versions of this product

Catalog No. AA14343A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1633-05-2
    Physical Form -325 Mesh, Typically 1 Micron Powder
    Assay Percent Range 99%
    Molecular Formula SrCO3
    Formula Weight 147.63
    MDL Number MFCD00011250
    Chemical Name or Material Strontium carbonate, 99%, Ba 1%

    Specifications

    Formula Weight 147.63
    Quantity 1kg
    EINECS Number 216-643-7
    Physical Form -325 Mesh, Typically 1 Micron Powder
    Density 3.7g/mL
    Assay Percent Range 99%
    Molecular Formula SrCO3
    MDL Number MFCD00011250
    CAS 1633-05-2
    Chemical Name or Material Strontium carbonate, 99%, Ba 1%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit