Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Strontium chloride, anhydrous, 95%

Manufacturer:  Alfa Aesar™ 1220236

Quantity 500g

 View more versions of this product

Catalog No. AA1220236


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10476-85-4
MFCD00011249
Strontium chloride, Anhydrous
158.53
14,8840

Specifications

10476-85-4
158.53
14,8840
Odorless
Hygroscopic
White
500g
95%
Powder
MFCD00011249
3.052g/mL
1.650
1250°C
873°C
Strontium chloride, Anhydrous

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : WK8400000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit