Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium chloride, anhydrous, 99.5% (metals basis)

Catalog No. p-4782162
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 10476-85-4 | Cl2Sr

Strontium chloride is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-85-4
Molecular Formula Cl2Sr
MDL Number MFCD00011249

Specifications

Boiling Point 1250°C
Melting Point 873°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.5%, metal basis
Solubility Information Freely soluble in water. Very slightly soluble in ethanol and acetone.
Formula Weight 158.53
Refractive Index 1.65
Sensitivity Hygroscopic
Density 3.052
Chemical Name or Material Strontium Chloride
Show More Show Less
Hazard Category H318-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection.
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 233-971-6
RTECSNumber WK8400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.