Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Strontium Chloride, Hexahydrate
SCI_ED

Catalog No. p-4416485 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Lab Grade
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

Specifications

Color White
Quantity 100 g
Assay Percent Range >99%
Linear Formula SrCl2·6H2O
Formula Weight 266.62
Grade Laboratory
Packaging Lab Grade
Density 1.9g/mL
Chemical Name or Material Strontium Chloride, Hexahydrate
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.