Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium chloride hexahydrate, 99%, extra pure

Catalog No. p-3527088
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle

CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

Specifications

Melting Point 115.0°C
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula SrCl2·6H2O
Merck Index 15, 8970
Solubility Information Solubility in water: 106.2 g/100mL (0°C). Other solubilities: soluble in alcohol
Formula Weight 266.62
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Strontium chloride hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.