Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium chloride, ultra dry, 99.995% (metals basis)

Catalog No. p-4407962
Click to view available options
Quantity:
5 g
25 g

CAS: 10476-85-4 | SrCl2

Strontium chloride  is used as a source of redness in fireworks. Strontium chloride is often used as a red coloring agent in pyrotechnics. It is employed in small quantities in glass-making and metallurgy. The radioactive isotope strontium-89, used for the treatment of bone cancer, is usually administered in the form of strontium chloride. Sea water aquaria require small amounts of strontium chloride, which is consumed in the production of the exoskeletons of certain plankton. SrCl2 is useful in reducing tooth sensitivity by forming a barrier over microscopic tubules in the dentin containing nerve endings that have become exposed by gum recession. strontium chloride exposure induces parthenogenetic activation of oocytes[6] which is used in developmental biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-85-4
Molecular Formula SrCl2
MDL Number MFCD00011249

Specifications

Boiling Point 1250°C
Melting Point 873°C
Physical Form Beads
Quantity 5 g
Assay Percent Range 99.995% (metals basis)
Solubility Information It is soluble in water,slightly soluble in ethanol,acetone. Insoluble in ammonia.
Formula Weight 158.53
Refractive Index 1.65
Sensitivity Hygroscopic
Density 3.052
Chemical Name or Material Strontium chloride
Show More Show Less
Hazard Category H318-H335
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 233-971-6
RTECSNumber WK8400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.