Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium chromate, 95%

Catalog No. p-4407963
Click to view available options
Quantity:
250 g
1 kg

CAS: 7789-06-2 | CrO4Sr | 203.61 g/mol

Strontium chromate is used an primer for zinc, magnesium, aluminum, and alloys used in aircraft manufacture. Used as a colorant in polyvinyl chloride resins. It is also used in aluminum flake coatings. paints, varnishes and oil colors. It is a used in water based wash primers, metal conditioners, additive to control the sulfate content of solutions in electrochemical processes. It is also used in paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-06-2
Molecular Formula CrO4Sr
Molecular Weight (g/mol) 203.61
MDL Number MFCD00049549
InChI Key NVKTUNLPFJHLCG-UHFFFAOYSA-N
Synonym strontium chromate, strontium yellow, strontium chromate vi, sutokuro t, deep lemon yellow, strontium chromate a, c.i. pigment yellow 32, strontium chromate 1:1, strontium chromate 12170
PubChem CID 24599
IUPAC Name strontium;dioxido(dioxo)chromium
SMILES [O-][Cr](=O)(=O)[O-].[Sr+2]

Specifications

Physical Form Powder
Quantity 250 g
Solubility Information Soluble in boiling water,dilute HCl,dilute CH3CO2H
Formula Weight 203.61
Density 3.895
Chemical Name or Material Strontium chromate
Hazard Category H302-H317-H330-H335-H340-H350-H361
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 232-142-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.