Strontium chromate, Alfa Aesar™

Manufacturer: Alfa Aesar 8902630

Quantity 250g

 View more versions of this product

Catalog No. AA8902630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-06-2
    Physical Form Powder
    Molecular Formula SrCrO4
    Formula Weight 203.61
    MDL Number MFCD00049549
    Chemical Name or Material Strontium chromate

    Specifications

    Formula Weight 203.61
    Quantity 250g
    EINECS Number 232-142-6
    Physical Form Powder
    Molecular Formula SrCrO4
    MDL Number MFCD00049549
    CAS 7789-06-2
    Chemical Name or Material Strontium chromate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit