Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium iodide, anhydrous, 99.99% (metals basis)

Catalog No. p-7058864
Click to view available options
Quantity:
5 g

CAS: 10476-86-5 | SrI2

Strontium iodide is used as a scintillation gamma radiation detector. It is also used in a range of hand-held radiation detection instruments as well as in medical, industrial and environmental applications. It is used in medicine as a substitute for potassium iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10476-86-5
Molecular Formula SrI2
MDL Number MFCD00049557

Specifications

Melting Point 515°C
Color White to Yellow
Physical Form Powder
Quantity 5 g
Merck Index 14,8844
Solubility Information Soluble in water and ethanol.
Formula Weight 341.43
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 4.550 g/mL
Assay (metals basis)
Chemical Name or Material Strontium iodide, Anhydrous
Show More Show Less
Hazard Category H290-H315-H319-H335
Hazard Statement GHS H Statement
H290-H335-H315-H319
May be corrosive to metals.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P234-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P390-P501c
EINECSNumber 233-972-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.