Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium isopropoxide, 96%

Catalog No. p-7049478
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 88863-33-6 | C6H14O2Sr | 205.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88863-33-6
Molecular Formula C6H14O2Sr
Molecular Weight (g/mol) 205.80
MDL Number MFCD00050487
InChI Key OHULXNKDWPTSBI-UHFFFAOYSA-N
Synonym strontium isopropoxide, strontium 2+ bis propan-2-olate, strontium propan-2-olate, diisopropoxystrontium, acmc-20egm0, strontium isopropylate, 2-propanol, strontiumsalt 9ci, strontium 2+ ion bis propan-2-olate
PubChem CID 6099237
SMILES [Sr++].CC(C)[O-].CC(C)[O-]

Specifications

Melting Point 130°C (decomposition)
Quantity 1 g
UN Number UN3205
Sensitivity Air and moisture sensitive
Solubility Information Soluble in isoprpanol.
IUPAC Name strontium(2+) bis(propan-2-olate)
Formula Weight 205.82
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Strontium isopropoxide
Hazard Category H228-H252-H314-H335
Hazard Statement GHS H Statement
H314-H318-H228-H252
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Precautionary Statement P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.