Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium nitrate, 99+%, ACS reagent

Catalog No. ac315171000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 570.0°C
Color White
pH 5.0 to 7.0 (5% soln. at 25°C)
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula Sr(NO3)2
Merck Index 15,8975
Solubility Information Solubility in water: 660g/L (20°C). Other solubilities: slightly soluble in alcohol and acetone
IUPAC Name strontium;dinitrate
Formula Weight 211.63
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 0.1% max. (105°C)
Packaging Plastic bottle
Density 2.99
Chemical Name or Material Strontium nitrate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
EINECSNumber 233-131-9
RTECSNumber WK9800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.