Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium nitrate, anhydrous, 98%

Catalog No. p-4904547
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol

In railroad flares, signal lights, marine signalsStrontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. Mixed with glycolic acid, it is used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
IUPAC Name strontium;dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 570°C
Color White
Physical Form Granules
Quantity 250 g
Assay Percent Range 98%
Solubility Information Freely soluble in water. Slightly soluble in alcohol and acetone.Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid.
Formula Weight 211.63
Odor Odorless
Sensitivity Hygroscopic
Density 2.99
Chemical Name or Material Strontium nitrate, Anhydrous
Show More Show Less
Hazard Category H272-H318
Precautionary Statement P210-P220-P221-P280-P305+P351+P338-P310-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 233-131-9
RTECSNumber WK9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.