Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium nitrate, anhydrous, 99.97% (metals basis)

Catalog No. p-7049868
Click to view available options
Quantity:
25 g
100 g

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol

Strontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. It mixed with glycolic acid and used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 570°C
Color White
Physical Form Crystalline
Quantity 25 g
Linear Formula Sr(NO3)2
UN Number UN1507
Merck Index 14,8845
Solubility Information Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid.
IUPAC Name strontium(2+) dinitrate
Formula Weight 211.63
Percent Purity 99.97%
Odor Odorless
Sensitivity Hygroscopic
Density 2.990 g/mL
Assay (metals basis)
Chemical Name or Material Strontium nitrate, Anhydrous
Show More Show Less
Hazard Category H272-H318
Hazard Statement GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P280-P305+P351+P338-P310-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: STRONTIUM NITRATE
EINECSNumber 233-131-9
RTECSNumber WK9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.