Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18614111 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-614-111

Supplier: Spectrum Chemical Manufacturing Corporation S168025GM

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10042-76-9

Specifications

pH 5.0 to 7.0
CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Loss on Drying 0.001
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.